Sodium hydroxide 10 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Caustic soda

5674-02EA 84.3 GBP
5674-02
Sodium hydroxide 10 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide

Caution: IMPORTANT: Material will freeze if stored below 5 °C. Protect from air to avoid absorption of carbon dioxide.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory, Research or Manufacturing Use Passes Test
Protect from air to avoid absorption of carbon dioxide. 9.9 - 10.1
IMPORTANT: Material will freeze if stored below 41 °F (5°C). = 2.0 %
Appearance (Clear, colorless liquid) Passes Test
Normality 9.9 - 10.1
Sodium Carbonate (Na₂CO₃) ≤2.0 %
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…
IMPORTANT: Material will freeze if stored below 41°F (5°C).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR