Medium pressure 4-port switching valves, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
V-101D V-101L V-101L-60 V-101T
554-3277EA 334 GBP
554-3277 554-3278 554-3279 554-3280
Medium pressure 4-port switching valves, Upchurch Scientific®
Valves Switching Valves
  • Biocompatible and chemically inert
  • Pressure rated to 500 psi (34 bar)