Reagent sprayer
Supplier: CAMAG
022.6100
552-0031EA 277 GBP
552-0031
Reagent sprayer
Atomisers
Glass atomiser. Cost efficient alternative for a TLC sprayer.
  • Can be used with compressed air or nitrogen
  • Reagents are atomised into a fine aerosol spray