GC capillary column splitters
Supplier: Trajan Scientific and Medical
18614600 123712 123711
548-1792EA 213 GBP
548-1792 548-1776 548-1775
GC capillary column splitters
Fittings and Connectors
Fixed outlet splitters for capillary columns
A fixed outlet splitter is recommended for splitting the flow from a capillary column between two detectors. The kit consists of a special Glass Lined Tubing (GLT™) splitter union, tubing and all ferrules required for installation.
Fixed inlet splitters for capillary columns
Where an injected sample is to be split between two capillary columns for simultaneous analysis, a fixed inlet splitter is recommended. The low internal volume, glass-lined design ensures an inert low dead volume connection. The two capillary columns used (usually differingMore Product Information