VWR®, Septa ND22
Supplier: VWR Collection
548-0159AEA 548 GBP
548-0159A 548-2179A 548-3338A 548-0616A 548-0615A 548-3232A
VWR®, Septa ND22
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.