VWR® ND18/ND22/ND28, Snap Cap Vials
Supplier: VWR Collection
548-0554EA 25.3 GBP
548-0554
VWR® ND18/ND22/ND28, Snap Cap Vials
Vials
These vials should only be used for storage of powders and solids.
  • Caps and vials can be ordered separately
  • Quick and easy to reopen and reseal
  • No liners are required in the cap