VWR®, Crimp Caps for Crimp Neck Vials ND13
Supplier: VWR Collection
548-1843AEA 80.9 GBP
548-1843A 548-0488A 548-0513A 548-0492A 548-1459A 548-0498A 548-2132A 548-0495A 548-0489A 548-2084A 548-1522A 548-0858A 548-0491A 548-0496A
VWR®, Crimp Caps for Crimp Neck Vials ND13
Closures Chromatography Vial Closures
Crimp Neck ND13 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Crimp top vials provide an excellent seal
  • Larger opening improves sample accessibility
ND13 vials with a crimp neck are available in clear version. For use in a broad range of instruments as GC and HPLC standard vials with a flat bottom. Broad variety of crimp caps mostly available with assembled stopper or septa are engineered to preventMore Product Information