Progesterone, Millipore®

Supplier: Merck Millipore Calbiochem

5341-25
5341-25EA 180 GBP
5341-25
Progesterone, Millipore®
Progesterone

⁴-Pregnene-3,20-dione


Steroid hormone secreted by the corpus luteum during the latter half of the menstrual cycle. Binds to the progesterone receptor in the nucleus to influence transcription.

Formula: C₂₁H₃₀O₂
MDL Number: MFCD00003658
CAS Number: 57-83-0
EINECS: 200-350-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR