Nickel(II) sulphate hexahydrate ≥99.97% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Nickelous sulphate, Hexahydrate, Nickel sulphate hexahydrate

53130.18 53130.30
53130.18EA 78.6 GBP
53130.18 53130.30
Nickel(II) sulphate hexahydrate ≥99.97% (metals basis)
Nickel sulphate hexahydrate

Purity: 99.97% min CAS No.: 10101-97-0 Molecular Formula: NiO4S.6H2O Formula Weight: 262.86 (154.77anhy) Physical Form: Lump Density: 2.070 Appearance: Green Odour: Odourless pH: 4.3-4.7 Merck Reference: 14,6517 UN No.: 3288 MDL No.: MFCD00149813

Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840
Density: 2.070
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1 III
Merck Index: 14,6517

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR