Hei-VAP Industrial Large Scale Rotary Evaporators
Supplier: Heidolph Instruments GmbH & Co.KG
518-12100-00 518-13100-00 518-14100-00 518-16100-00 518-18100-00 518-19100-00 518-18102-00 518-19102-00
531-3412EA 0 GBP
531-3412 531-3413 531-3414 531-3416 531-3108 531-3109 HEID518-18102-00 HEID518-19102-00
Hei-VAP Industrial Large Scale Rotary Evaporators
Evaporators Rotary Evaporators
Hei-VAP Industrial large scale rotary evaporators for enhanced vacuum control standards and for maximum safety.
  • Excellent user safety with high-impact transparent PMMA door
  • Non-fogging safety glass and metal frame guard hood provides excellent user protection
  • The evaporation flask is illuminated during operation for increased visibility
  • Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180 °C
  • Receiver cassettes and additional PMMA door housing protect against threat of glassware breakage
  • Comes as
More Product Information