VWR®, Centrifuge Tube Stands
Supplier: VWR Collection
525-1003EA 14.2 GBP
525-1003
VWR®, Centrifuge Tube Stands
Racks Tube Racks
PP, green.
  • Free from DNase and RNase and non pyrogenic
  • Autoclavable at 121 °C and freezable to –80 °C
  • Very good chemical resistance
Suitable for 1,5/2,0 ml microcentrifuge tubes and 15/50 ml conical centrifuge tubes.