VWR®, Centrifuge Tubes, Light-Sensitive
Supplier: VWR Collection
525-0603EA 149 GBP
525-0603 525-0598 525-0599 525-0600 525-0601 525-0602
VWR®, Centrifuge Tubes, Light-Sensitive
Tubes Centrifuge Tubes
PP, high grade gamma resistant, conical, amber, with white HDPE plug style cap with deep sealing area.
  • Tubes and caps are autoclavable and freezable
  • Gamma irradiated
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Longer length screw caps with sealing ring prevent any leakage
  • Available in 15 or 50 ml sizes and offered in a variety of packaging formats
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Sustainable design based on virgin resins
  • Blocks 99% of light with wavelength less than 550
More Product Information