Multifuge™ X4/X4R Pro Centrifuges (IVD-Marked)
Supplier: Thermo Scientific
75009500 75009515 75016057 75016058 75016081
521-2724EA 5229 GBP
521-2724 521-2726 521-2732 521-2733 521-2741
Multifuge™ X4/X4R Pro Centrifuges (IVD-Marked)
Centrifuges Benchtop Centrifuges
Process a range of general-purpose applications including cell culture, microplate, bioproduction, blood separation, and microbiology with the Thermo Scientific™ Multifuge™ X4 Pro centrifuge series featuring new touch screen technology, making daily tasks easier.
  • Process up to 40×50 ml conical tubes, 196×5/7 ml blood tubes, 24 micro plates
  • Process clinical protocols, cell culture applications, and microplate processing
  • Modern, intuitive touchscreen interface enables easy access and programming for faster results
  • Lighthouse mode: Easily monitor run progress
More Product Information