Microcentrifuges, mySPIN™
Supplier: Thermo Scientific
75004061 75004081
521-2155EA 377 GBP
521-2155 521-2156
Microcentrifuges, mySPIN™
Centrifuges Microcentrifuges
The mySPIN™ centrifuges are available as a six-place version with fixed speed and as a 12-place version with variable speed setting. Both models feature an ergonomic design and a compact footprint.
  • Quiet operation, low vibration
  • Tool-free, quick rotor exchange
  • MySPIN™ six unit with palm-shaped lid for left- or right-handed users; unit will only start to operate when lid is closed
  • MySPIN™ 12 with imbalance/tilt safety cut-out, back-lit display (can be set for rpm or RCF) and safety interlock
Core applications are the purification of DNA, RNA, proteinMore Product Information