Rotors for Centrifuge 5430/5430 R
Supplier: EPPENDORF
5427705.007 5427710.000 5427712.003 5427714.006 5427700.005 5427716.009 5427754.008 5427750.002 5427753.001 5427755.004 5427757.007 5427752.005
521-2013EA 478 GBP
521-2013 521-2039 521-2008 521-2016 521-2010 521-2017 521-1940 521-1941 521-1741 521-2067 521-2068 521-1740
Rotors for Centrifuge 5430/5430 R
Centrifuges Rotors
Eppendorf rotors for centrifuge 5430/5430 R, precision at its core. Maximise your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight and PTFE coated options available
Explore enhanced centrifugation possibilitiesMore Product Information