Titration solvent ASTM D-2710 / D-1159, VOLUSOL®

Supplier: VWR Chemicals
Danger

5203.2500EA 119 GBP
5203.2500
Titration solvent ASTM D-2710 / D-1159, VOLUSOL®
Titration solvent

Acetic acid/ methanol/ dichloromethane/ sulphuric acid/ water used in the measurement of unsaturated hydrocarbons in petroleum distillates (71.4/13.5/13.5/0.3/1.6 v/v).

Density: 1.029
Storage Temperature: Ambient
UN: 2920
ADR: 8 (3) II

Order Now

Specification Test Results

Density (20/4) 1.05 - 1.07
Identification Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR