Filter plates, 96-well, for ultrafiltration, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
8034 8036 8035 8033 8166 8165 8164 8163
518-0032EA 426 GBP
518-0032 518-0029 518-0028 518-0027 738-0126 738-0125 738-0124 738-0123
Filter plates, 96-well, for ultrafiltration, AcroPrep™ Advance
Microplates Filter Plates
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications, gross fractionation and lysate clarification and general filtration.
  • High biomolecule recovery - Omega® membrane typically results in ≥90% recovery of target biomolecules
  • Well
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable