Vacuum filtration manifolds, Microsart®
Supplier: Sartorius
168M1-MS 168M2-MS 168M3-MS 168M6-MS 168M1-BS100 168M2-BS100 168M3-BS100 168M6-BS100 168M1-BS250 168M3-BS250 168M6-BS250 168M1-SS100 168M3-SS100 168M6-SS100 168M1-SS500 168M3-SS500 168M6-SS500
516-1086EA 1760 GBP
516-1086 516-1087 516-1088 516-1089 516-1090 516-1091 516-1092 516-1093 516-1094 516-1095 516-1096 516-1097 516-1098 516-1099 516-1100 516-1151 516-1156
Vacuum filtration manifolds, Microsart®
Manifolds
Microsart® Manifolds accurately detect and quantify microorganisms in liquid samples. Simply select the proper manifold for your individual microbiological testing procedure, choose between reusable and single-use funnels or filtration units, and then decide between different sizes of manifolds and enjoy the manifold that adapts to your needs.
  • 100% stainless steel for minimised cleaning effort and full autoclavability without taking apart
  • Sterile venting in critical applications reduces risk of secondary contamination
  • Quick connection adapters for fast
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable