Capsule filters, Polycap™ AS, Whatman™
Supplier: Whatman products (Cytiva)
6705-3604 6706-3602 6709-3602 6705-3602 2707NS 6708-3602 2706T 2606T
516-4436EA 59.8 GBP
516-4436 516-4442 516-4445 515-0108 516-0966 516-0443 516-0965 516-0957
Capsule filters, Polycap™ AS, Whatman™
Filters Membrane Filter Capsules
These Polycap™ AS (aqueous solutions) capsule filters combine a glass microfiber (GMF) prefilter over a nylon membrane final filter to provide maximum filtration of difficult, particulate-laden solutions.
  • First layer (GMF) acts as a prefilter to ensure longer membrane life and higher filtration efficiency
  • Nylon membrane filter layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates
  • PP housing is thermally fused (no glues, adhesives, or extraneous materials), reducing extractable levels
  • Can be tested with bubble point,
More Product Information