Glass microfibre filters, 934-AH™ RTU, Whatman™
Supplier: Whatman products (Cytiva)
9907-042 9907-055 9907-047 9907-090 9907-9436 2827-047 3827-035 3827-042 3827-047 3827-070 3827-090 4827-035 4827-042 4827-047 4827-070 4827-090 9907-070 9927-047 9927-070 9927-090
513-0114EA 103 GBP
513-0114 513-0116 513-0115 513-0117 WHAT9907-9436 513-0140 513-0127 513-0128 513-0129 513-0130 513-0131 513-0132 513-0133 513-0134 513-0135 513-0136 WHAT9907-070 513-0137 513-0138 513-0139
Glass microfibre filters, 934-AH™ RTU, Whatman™
Filters Glass Fibre Filters
These 934-AH™ RTU grade filter papers have been pre-washed, dried (or ignited), cooled, desiccated and weighed, which eliminates the pre-treating steps that are otherwise required by the US EPA Laboratory Standard Method 2540 parts C, D, and E. These filter papers are widely used for dissolved, suspended and volatiles solid analysis. They are also used as a wastewater filter for a wide range of water monitoring applications including the monitoring of rivers, lakes and coastal waters, as well as analysing purified discharge of wastewater treatment plants and monitoring discharge water fromMore Product Information