Sealing film and foil for microplates, Whatman™
Supplier: Whatman products (Cytiva)
7704-0001
512-1087EA 79.3 GBP
512-1087
Sealing film and foil for microplates, Whatman™
Seals Microplate Seals
Seals are used to control humidity and reduce evaporation of samples. They prevent spills and contamination.
  • Cold seals in clear polyester or PP are self-sticking with inert adhesive
  • Heat seals are suitable for PP microplates only and are applied using heat and pressure
Order Now
SPECIFICATIONS
Environmentally Preferable