Filter funnel heads
Supplier: DWK Life Sciences
247202407 247205001 247209003 247212408 247215002 247219004
511-5110EA 33.1 GBP
511-5110 511-5120 511-5130 511-5117 511-5127 511-5137
Filter funnel heads
Funnels Filtration Funnels Filter Funnels
Filter head in DURAN® borosilicate glass 3.3, clear, PP funnel and two fluorinated rubber sealing rings.
  • Easily interchangeable discs
  • Safe and simple removal of filtered material
  • Discs easily cleaned on both sides
Slit sieve 511-5126 (48 mm Ø) suitable as paper filter or membrane support on the 250 ml model.
Ordering information: Threaded filter head with funnel and two sealing rings, without filter disc.
Order Now
SPECIFICATIONS
Environmentally Preferable