Filtering beakers, DURAN®
Supplier: DWK Life Sciences
211312409 211312906 211313602 211314401 211315406 211316308 211316805 211317304 211318609 211318806 211319108
511-0315EA 35.5 GBP
511-0315 511-0316 511-0340 511-0317 511-0318 511-0319 511-0320 511-0321 511-0322 511-0323 511-0324
Filtering beakers, DURAN®
Beakers
DURAN®, borosilicate glass 3.3, clear.
  • Increased wall thickness for better mechanical properties
  • Limited use as cooking glass due to reduced thermal shock resistance
  • Available graduated or non graduated
Thick walled, with spout, easy to read scale and large labelling field.
Above 10000 ml without graduation