VWR® Ultrasonic Sieving System
Supplier: HAVERBOECKER
510-1274EA 4530 GBP
510-1274 510-1282 510-1276 510-1275
VWR® Ultrasonic Sieving System
Shakers Sieve Shakers
Using ultrasonic frequency variation for test sieving, even powders with separating cuts of ≤300 micron can be sieved. Fitting at least one HAVER-test sieve with a frequency variation system enables such analyses.
The ultrasonic frequency variation system consists of a clamping ring, which is connected by a converter to a generator. The generator converts electrical energy into high frequency energy, which is then converted to mechanical energy by the converter and transferred to the clamping ring. Sound waves cause the test sieve frame to oscillate at high frequency. These oscillationsMore Product Information