Formic acid 98 - 100% 98-100, LiChropur™ LC-MS, Supelco®

Supplier: Merck
Danger

Synonyms: Aminic Acid, Methanoic acid

5.33002.0050
5.33002.0050EA 122 GBP
5.33002.0050
Formic acid 98 - 100% 98-100, LiChropur™ LC-MS, Supelco®
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Density: 1.22
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3) II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR