VWR®, Automated Cell Counters
Supplier: VWR Collection
49862-2000EA 3510 GBP
49862-2000 49893-2000 49893-4950 49893-4951 49893-4952
VWR®, Automated Cell Counters
Cell Counters and Flow Cytometers Automated Cell Counters
Save time and reduce variability with VWR® automated cell counter and VWR® automated cell counter Fluo. Intelligent image recognition and advanced optical imaging technology to achieve accurate and automatic cell analysis, cell concentration and cell size determination.
  • Wide application: cell counting, cell viability, AO/PI staining, cell transfection efficiency, cell expression
  • All-in-one machine with a compact design
  • Dual channel option in VWR® automated cell counter Fluo
  • 3 interchangeable fluorescent cubes (green, blue, red)
  • Cell counting slide
More Product Information