Gamma/beta ray sterilisation indicators
Supplier: SÜDPACK MEDICA
7600018 7600016 7600015
489-0004EA 260 GBP
489-0004 489-0005 489-0099
Gamma/beta ray sterilisation indicators
Sterilisation Indicators
The discs are available in a variety of blank or printed versions.
The 489-0004 indicator carries the text: 'red is gamma exposed'.
The 489-0099 and 489-0005 indicators turn from yellow to red after exposure to gamma/beta rays.
Packaging: Roll of 5000 labels.
Order Now
SPECIFICATIONS
Environmentally Preferable