VWR®, Cryogenic Vials
Supplier: VWR Collection
479-1208EA 0 GBP
479-1208 479-1218 479-1237 479-1238
VWR®, Cryogenic Vials
Vials
Designed for the storage of biological material, human or animal cells, at temperatures as low as –196 °C (should only be used in the gas phase of liquid nitrogen).
  • Graduations and white marking area
  • Sterilised by gamma radiation
  • Compatible with most storage systems
Closures and tubes are made of PP, and both have the same coefficient of expansion, further enhancing the leakproof qualities of these vials at various temperatures. Round bottom vials can be centrifuged up to 17000×g.
Please note that 479-1237 has a PP screw cap and there are no capMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable