Viscosity standard, Specpure®, Nominal Values at 40 and 100 °C

Supplier: Thermo Fisher Scientific

45340.AP 47036.AP 47100.AP 47155.AP 47158.AP
45340.APEA 102 GBP
45340.AP 47036.AP 47100.AP 47155.AP 47158.AP
Viscosity standard, Specpure®, Nominal Values at 40 and 100 °C
Standards Viscometry Standards

Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00084411

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00084411 MFCD00084411
Physical form Liquid Liquid
Odour/scent Odourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR