Sulphamethoxypyridazine, Supelco®

Supplier: Merck

Synonyms: 4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide

MQ200

46858-250MG-R
46858-250MG-REA 51.7 GBP
46858-250MG-R
Sulphamethoxypyridazine, Supelco®
Sulphamethoxypyridazine
Formula: C₁₁H₁₂N₄O₃S
MW: 280.3 g/mol
MDL Number: MFCD00057372
CAS Number: 80-35-3
EINECS: 201-272-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3c3118107c95e79b2ad217431f1f337df1cdf61&vid=37908fa89983bc19dc4fa3a629540bc35236abf0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR