Diaion® SA10A (Cl) 1,3 meq/ml on Poly(styrene-divinylbenzene), cross-linked, beads 300 - 1180 µm, gel type_strongly basic resin_ion-exchange
Supplier: Thermo Fisher Scientific
46796.A1
46796.A1EA 77.3 GBP
46796.A1
Diaion® SA10A (Cl) 1,3 meq/ml on Poly(styrene-divinylbenzene), cross-linked, beads 300 - 1180 µm, gel type_strongly basic resin_ion-exchange
Diaion® SA10A (Cl)
Diaion® SA10A(Cl) is an ion exchange resin, cross-linked, strongly basic gel type I, 1.3 meq/ml on PS-DVB.
Used in the waste water treatment, demineralisation, decolorisation, metal recovery and high purity water.