Desiccator cabinets, SICCO Star series
Supplier: Bohlender
V1871-07 V1875-07 V1879-07 V1841-07
467-4126EA 1180 GBP
467-4126 467-0053 467-0114 467-0112
Desiccator cabinets, SICCO Star series
Desiccators Desiccator Cabinets
These transparent PMMA or borosilicate 3.3 glass cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed for storing or drying humidity-sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA samples. Super-Star cabinets can be filled with inert gas, e.g. nitrogen to allow safe and secure storage of poisonous chemicals.
  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable