Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M aqueous solution Indicator_Redox, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate

46270-250ML 46270-50ML
46270-250MLEA 127 GBP
46270-250ML 46270-50ML
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M aqueous solution Indicator_Redox, Fluka™
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)
Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Boiling Pt: 100 °C (solvent)
Melting Pt: 0 °C
Density: 1.006 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00036428
CAS Number: 14634-91-4

Order Now

Specification Test Results

Appearance of the substance complying
Density (D 20/4) 1.003 - 1.008
Suitability as indicator complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR