Baths for immersion thermostats
Supplier: VWR Collection
462-0263EA 1190 GBP
462-0263 462-0264 462-0265 462-0267 462-0268 462-0270 462-0269
Baths for immersion thermostats
Baths Water Baths, Unstirred
Made from stainless steel or polycarbonate. Ideal for use with VWR® immersion thermostats. For use where the durability of stainless steel or the transparency of polycarbonate are advantageous.
  • Open baths, ideal for use with immersion thermostats from VWR® or other manufacturers
  • Available in stainless steel or polycarbonate
  • Robust and durable
  • Easy to clean
Note: The temperature ranges listed below apply only when the vessels are used with immersion thermostats; an immersion thermostat is required but not supplied with the bath.
Order Now
SPECIFICATIONS
Environmentally Preferable