VWR® ARISTAR®, Hafnium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
85569.180EA 215 GBP
85569.180 457292B
VWR® ARISTAR®, Hafnium Standard Solutions for ICP-MS
Standards Cationic Standards Hafnium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities