VWR®, Scandium standard solution, 1000 mg/L Sc in dil. nitric acid (from Sc2O3) ARISTAR® standard for ICP
Supplier: VWR Chemicals
455964UEA 695 GBP
455964U 455962S
VWR®, Scandium standard solution, 1000 mg/L Sc in dil. nitric acid (from Sc2O3) ARISTAR® standard for ICP
Standards Cationic Standards Scandium Standards
Sc₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable