Bis(tri-tert-butylphosphine)palladium(0)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)

Inert_Gas_filled

44845.03 44845.01
44845.03EA 248 GBP
44845.03 44845.01
Bis(tri-tert-butylphosphine)palladium(0)
Bis(tri-tert-butylphosphine)palladium(0)

CAS No.: 53199-31-8 Molecular Formula: C24H54P2Pd Formula Weight: 511.06 Storage Temperature: -30°C to -10°C Physical Form: Powder, packaged under inert atmosphere Sensitivity: Light, Air and Moisture Sensitive Appearance: White to off-white Melting Point: 258-272° MDL No.: MFCD03094580


Coupling reactions, Heck couplings

Formula: C₂₄H₅₄P₂Pd
MW: 511.05 g/mol
Melting Pt: 258…272 °C
Storage Temperature: Freezer
MDL Number: MFCD03094580
CAS Number: 53199-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR