tert-Butyl ethyl ether standard_analytical, Supelco®

Supplier: Merck

Synonyms: 2-Ethoxy-2-methylpropane

442795
442795.EA 182 GBP
442795.
tert-Butyl ethyl ether standard_analytical, Supelco®
tert-Butyl ethyl ether
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 71
Melting Pt: –97 °C
Density: 0.752 g/cm³ (20 °C)
MDL Number: MFCD00009225
CAS Number: 637-92-3
EINECS: 211-309-7
UN: 1179
ADR: 3 II
Merck Index: 13,03809
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927d89821a52dcb33998d5099e63cc54c7f276c8&vid=927d89821a52dcb33998d5099e63cc54c7f276c8&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR