Squalene standard_analytical, Supelco®

Supplier: Merck

Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene

442785
442785.EA 425 GBP
442785.
Squalene standard_analytical, Supelco®
Squalene
Formula: C₃₀H₅₀
MW: 410.73 g/mol
Boiling Pt: 429
Melting Pt: –5 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 200
MDL Number: MFCD00008912
CAS Number: 111-02-4
EINECS: 203-826-1
Merck Index: 13,08847
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d47752616c4ee55d8684f4da254afd15d45eb6c7&vid=d47752616c4ee55d8684f4da254afd15d45eb6c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR