Magnetic stirring bar retrievers, RSE, PTFE
Supplier: IKA
1293100
442-0902EA 124 GBP
442-0902
Magnetic stirring bar retrievers, RSE, PTFE
Stirrers Stirring Tools
These PTFE-coated retriever used for removing all stirring bars upto 80 mm in length.
  • Weight: 0,048 kg