N-Methyl-N-(trimethylsilyl)trifluoroacetamide, Supelco®

Supplier: Merck
Danger

Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide

MQ100

44156-5ML-F
44156-5ML-FEA 184 GBP
44156-5ML-F
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, Supelco®
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formula: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Boiling Pt: 130
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR