Pyridine, anhydrous 99,5

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Azabenzene, Azine

Argon_Chemseal

43799.AE 43799.K2
43799.AEEA 72.2 GBP
43799.AE 43799.K2
Pyridine, anhydrous 99,5
Pyridine

Purity: 99.5+% CAS No.: 110-86-1 Molecular Formula: C5H5N Formula Weight: 79.10 Flash Point: 20°(68°F) Physical Form: Liquid Density: 0.978 Refractive Index: 1.5100 Odour: Unpleasant Melting Point: -42° Boiling Point: 115° Beilstein Reference: 103233 Merck Reference: 14,7970 UN No.: 1282 MDL No.: MFCD00011732

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C
Melting Pt: –42 °C
Density: 0.978
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 14,7970
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR