Dimethyldichlorosilane 2% heptane, Electran Electrophoresis

Supplier: VWR Chemicals
Danger

Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane

437494JEA 104 GBP
437494J
Dimethyldichlorosilane 2% heptane, Electran Electrophoresis
Dimethyldichlorosilane

Designed to give water repellant properties to glassware. Suitable for use in electrophoresis to prevent adhesion of gels to glass plates.

Formula: C₂H₆Cl₂Si
MW: 129.06 g/mol
Boiling Pt: 70
Melting Pt: -76
Flash Pt: -16
MDL Number: MFCD00000491
CAS Number: 75-78-5
EINECS: 200-901-0
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR