Fluorescamine

Supplier: Thermo Fisher Scientific

Synonyms: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione

43749.MC 43749.MB 43749.ME
43749.MCEA 87.4 GBP
43749.MC 43749.MB 43749.ME
Fluorescamine
Fluorescamine

CAS No.: 38183-12-9 Molecular Formula: C17H10O4 Formula Weight: 278.27 Physical Form: Powder Appearance: White to cream Odour: Odourless Melting Point: 155-158° MDL No.: MFCD00005928


Applications: For fluorescent detection of amino acids, peptides and proteins

Formula: C₁₇H₁₀O₄
MW: 278.26 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00005928
CAS Number: 38183-12-9
EINECS: 253-814-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR