n-Nonane, anhydrous 99%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

435631000
435631000.EA 243 GBP
435631000.
n-Nonane, anhydrous 99%, AcroSeal®
n-Nonane
Formula: H₃C(CH₂)₇CH₃
MW: 128.26 g/mol
Boiling Pt: 151 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.718 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00009574
CAS Number: 111-84-2
UN: 1920
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
Water ≤0.005 %
GC ≥98.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR