n-Dodecane, anhydrous 99%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dodecane

434590010 434591000
434590010.EA 988 GBP
434590010. 434591000.
n-Dodecane, anhydrous 99%, AcroSeal®
n-Dodecane
Formula: H₃C(CH₂)₁₀CH₃
MW: 170.34 g/mol
Boiling Pt: 215…217 °C (760 mmHg)
Melting Pt: –12 °C
Density: 0.753 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008969
CAS Number: 112-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d3d75a63f52fbe2a8be1681d9233be6dc0928&vid=3268fe23453ad703df2782bd3c7ba6b939ba00cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR