Palladium 5% on calcium carbonate for use as a catalyst lead poisoned Type A306060-5

Supplier: Thermo Fisher Scientific
Danger

43172.14 43172.06 43172.22
43172.14EA 369 GBP
43172.14 43172.06 43172.22
Palladium 5% on calcium carbonate for use as a catalyst lead poisoned Type A306060-5
Palladium

CAS No.: 7440-05-3 Physical Form: Powder UN No.: UN3077 MDL No.: MFCD03427452


Applications: Selective hydrogenation

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR