Zirconium standard dissolved in oil, 1000 µg/g Zr in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific

43127.18
43127.18EA 62.3 GBP
43127.18
Zirconium standard dissolved in oil, 1000 µg/g Zr in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Zirconium Standards

Physical form: liquid
MDL No.: MFCD00011303

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00011303

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR