Succinyl chloride ∼95%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Succinyl dichloride

430801000
430801000.EA 108 GBP
430801000.
Succinyl chloride ∼95%, AcroSeal®
Succinyl chloride
Formula: C₄H₄Cl₂O₂
MW: 154.98 g/mol
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: 16…17 °C
Density: 1.407 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000749
CAS Number: 543-20-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4720 (20°C, 589 nm)
Titration RCOCl ≥94 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR