Trimethyltin chloride 20% w/w 1 M THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Chlorotrimethylstannane

427851000
427851000.EA 319 GBP
427851000.
Trimethyltin chloride 20% w/w 1 M THF, AcroSeal®
Trimethyltin chloride
Formula: C₃H₉ClSn
MW: 199.27 g/mol
Boiling Pt: 96
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000520
CAS Number: 1066-45-1
UN: 1992
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.90 to 1.10 M
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR